Product Details

SNP ID
rs139439104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26188876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCGTGTAGGTGACAGCATCGCG[A/G]ATTACATTTTCCAGGAAAACTTTCA
Phenotype
MIM: 602792 MIM: 602805 MIM: 602811 MIM: 602823
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AD PubMed Links

Gene Details

Gene
HIST1H2AD
Gene Name
histone cluster 1, H2ad
There are no transcripts associated with this gene.

Gene
HIST1H2BE
Gene Name
histone cluster 1, H2be
There are no transcripts associated with this gene.

Gene
HIST1H3D
Gene Name
histone cluster 1, H3d
There are no transcripts associated with this gene.

Gene
HIST1H4D
Gene Name
histone cluster 1, H4d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003539.3 201 Silent Mutation ATC,ATT I67I NP_003530.1

View Full Product Details