Product Details

SNP ID
rs139560681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89645079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATATTTGCATTCCGGGCAGCCTGA[A/T]CTGCTGCCAGGACTCTTTCTTTGCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929057 PubMed Links

Gene Details

Gene
LOC101929057
Gene Name
uncharacterized LOC101929057
There are no transcripts associated with this gene.

Gene
LYRM2
Gene Name
LYR motif containing 2
There are no transcripts associated with this gene.

Gene
MDN1
Gene Name
midasin AAA ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014611.2 16530 Missense Mutation GAT,GTT D5513V NP_055426.1
XM_005248700.3 16530 Missense Mutation GAT,GTT D3508V XP_005248757.1
XM_006715405.3 16530 Missense Mutation GAT,GTT D3661V XP_006715468.1
XM_011535635.2 16530 Missense Mutation GAT,GTT D5416V XP_011533937.1
XM_011535636.2 16530 Intron XP_011533938.1

View Full Product Details