Product Details
- SNP ID
-
rs139620979
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:52187660 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGAAGGCAGGAATCACAATCCCA[C/T]GAAATCCAGGATGCCCAAATTCTGA
- Phenotype
-
MIM: 603149
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL17A
PubMed Links
Gene Details
- Gene
- IL17A
- Gene Name
- interleukin 17A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002190.2 |
130 |
Missense Mutation |
CGA,TGA |
R29* |
NP_002181.1 |
View Full Product Details