Product Details

SNP ID
rs139861886
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31529903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAAGCTGAAGGAACAGATGAGG[A/C]GGCACCAAGAGAGCCTTGGAGGAGG
Phenotype
MIM: 142560 MIM: 609624
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ATP6V1G2-DDX39B PubMed Links
Additional Information
For this assay, SNP(s) [rs3093979] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ATP6V1G2-DDX39B
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
DDX39B
Gene Name
DEAD-box helicase 39B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004640.6 381 Intron NP_004631.1
NM_080598.5 381 Intron NP_542165.1
Gene
MCCD1
Gene Name
mitochondrial coiled-coil domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011700.2 381 Silent Mutation AGG,CGG R110R NP_001011700.2
Gene
SNORD117
Gene Name
small nucleolar RNA, C/D box 117
There are no transcripts associated with this gene.

View Full Product Details