Product Details

SNP ID
rs140038883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27132789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAAGCGCCAACGAAAAGGAAAA[A/T]CAGCGTGAGCAGGGTATGACAAGGC
Phenotype
MIM: 615012 MIM: 615044 MIM: 615045 MIM: 602833
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2AG PubMed Links

Gene Details

Gene
HIST1H2AG
Gene Name
histone cluster 1, H2ag
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021064.4 8 Intron NP_066408.1
Gene
HIST1H2BJ
Gene Name
histone cluster 1, H2bj
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021058.3 8 UTR 5 NP_066402.2
Gene
HIST1H2BK
Gene Name
histone cluster 1, H2bk
There are no transcripts associated with this gene.

Gene
HIST1H4I
Gene Name
histone cluster 1, H4i
There are no transcripts associated with this gene.

View Full Product Details