Product Details

SNP ID
rs140124183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73681161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTAAATCCATGGCTTTGTTCT[A/G]TTCTCTCTCTTTTTTACCCCAGTGA
Phenotype
MIM: 608859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD109 PubMed Links

Gene Details

Gene
CD109
Gene Name
CD109 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159587.2 Intron NP_001153059.1
NM_001159588.2 Intron NP_001153060.1
NM_133493.4 Intron NP_598000.2
XM_005248659.3 Intron XP_005248716.1
XM_011535473.1 Intron XP_011533775.1
XM_017010283.1 Intron XP_016865772.1

View Full Product Details