Product Details
- SNP ID
-
rs140140644
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:108858200 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCTGCACCATCTTTTATGCTAGC[A/C]TCCATGCATCCAGTTTTGAGTCATC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARMC2
PubMed Links
Gene Details
- Gene
- ARMC2
- Gene Name
- armadillo repeat containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286609.1 |
388 |
Intron |
|
|
NP_001273538.1 |
NM_032131.5 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
NP_115507.4 |
XM_005267154.3 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_005267211.1 |
XM_005267155.2 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_005267212.1 |
XM_005267157.3 |
388 |
UTR 5 |
|
|
XP_005267214.1 |
XM_006715573.2 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_006715636.1 |
XM_006715574.2 |
388 |
UTR 5 |
|
|
XP_006715637.1 |
XM_011536166.1 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_011534468.1 |
XM_011536167.1 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_011534469.1 |
XM_011536168.2 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_011534470.1 |
XM_011536170.2 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_011534472.1 |
XM_011536172.2 |
388 |
Intron |
|
|
XP_011534474.1 |
XM_011536176.2 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_011534478.1 |
XM_017011350.1 |
388 |
UTR 5 |
|
|
XP_016866839.1 |
XM_017011351.1 |
388 |
Missense Mutation |
ATC,CTC |
I74L |
XP_016866840.1 |
XM_017011352.1 |
388 |
UTR 5 |
|
|
XP_016866841.1 |
View Full Product Details