Product Details

SNP ID
rs140176422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132570714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCTTCATCTCCATCGACAGGTA[C/T]ATTGCGGTTACTGACCCCCTGGTCT
Phenotype
MIM: 608923
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAAR6 PubMed Links

Gene Details

Gene
TAAR6
Gene Name
trace amine associated receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175067.1 393 Silent Mutation TAC,TAT Y131Y NP_778237.1

View Full Product Details