Product Details

SNP ID
rs140177705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:139373747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCATCGCATGCCTGATGCCGCT[C/G]GTGGCATTCATGTTGCCCGCGCCGT
Phenotype
MIM: 602937
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CITED2 PubMed Links

Gene Details

Gene
CITED2
Gene Name
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168388.2 444 Silent Mutation ACC,ACG T66T NP_001161860.1
NM_001168389.2 444 Silent Mutation ACC,ACG T71T NP_001161861.2
NM_006079.4 444 Silent Mutation ACC,ACG T66T NP_006070.2

View Full Product Details