Product Details

SNP ID
rs140192335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30546174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATCCCCTTCTCCCTCCTCATC[C/T]GCATCCCGGTCCTCCTCTCCCTCCT
Phenotype
MIM: 143024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNL1 PubMed Links

Gene Details

Gene
GNL1
Gene Name
G protein nucleolar 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005275.3 3113 Silent Mutation GCA,GCG A574A NP_005266.2
XM_005249015.3 3113 Silent Mutation GCA,GCG A574A XP_005249072.1

View Full Product Details