Product Details
- SNP ID
-
rs140231408
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:21594616 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCGGACTCGGGCGCCGGCCTCGAG[C/G]TGGGAATCGCCTCCTCCCCCACGCC
- Phenotype
-
MIM: 184430
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SOX4
PubMed Links
Gene Details
- Gene
- SOX4
- Gene Name
- SRY-box 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003107.2 |
876 |
Missense Mutation |
CTG,GTG |
L28V |
NP_003098.1 |
View Full Product Details