Product Details

SNP ID
rs140243222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143495749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTAGAGCCCAGCCCCATTTACA[C/G]GGCATCTGATGAATGGTTAGCTGTG
Phenotype
MIM: 136820 MIM: 603164
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FUCA2 PubMed Links

Gene Details

Gene
FUCA2
Gene Name
fucosidase, alpha-L- 2, plasma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032020.4 1611 Silent Mutation CCC,CCG P454P NP_114409.2
Gene
PEX3
Gene Name
peroxisomal biogenesis factor 3
There are no transcripts associated with this gene.

View Full Product Details