Product Details

SNP ID
rs140265864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110959184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTCACCAGGAGCAGTTGGTT[C/T]TGGTGGAATTATCAGGAATTATTGA
Phenotype
MIM: 611784
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF3C6 PubMed Links

Gene Details

Gene
GTF3C6
Gene Name
general transcription factor IIIC subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138408.3 Intron NP_612417.1

View Full Product Details