Product Details
- SNP ID
-
rs140276665
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:28245289 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAAACTTCTCTCACACTCATTAC[A/G]TTTATAAGACACGGGAGCCTCAGTA
- Phenotype
-
MIM: 611643
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZKSCAN4
PubMed Links
Gene Details
- Gene
- ZKSCAN4
- Gene Name
- zinc finger with KRAB and SCAN domains 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001304506.1 |
2476 |
Missense Mutation |
CGT,TGT |
R334C |
NP_001291435.1 |
NM_019110.4 |
2476 |
Missense Mutation |
CGT,TGT |
R489C |
NP_061983.2 |
XM_005249095.3 |
2476 |
Missense Mutation |
CGT,TGT |
R489C |
XP_005249152.1 |
XM_005249097.3 |
2476 |
Missense Mutation |
CGT,TGT |
R386C |
XP_005249154.1 |
XM_005249098.3 |
2476 |
Missense Mutation |
CGT,TGT |
R386C |
XP_005249155.1 |
XM_005249100.4 |
2476 |
Missense Mutation |
CGT,TGT |
R362C |
XP_005249157.2 |
XM_006715095.3 |
2476 |
Missense Mutation |
CGT,TGT |
R352C |
XP_006715158.1 |
XM_011514588.2 |
2476 |
Missense Mutation |
CGT,TGT |
R460C |
XP_011512890.1 |
XM_011514589.2 |
2476 |
Missense Mutation |
CGT,TGT |
R334C |
XP_011512891.1 |
XM_017010844.1 |
2476 |
Missense Mutation |
CGT,TGT |
R334C |
XP_016866333.1 |
View Full Product Details