Product Details
- SNP ID
-
rs140448519
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:73516143 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACTTGATACCTCAGAGCACTGGC[A/T]GATGGGAAGGCATTTTATCTAATTC
- Phenotype
-
MIM: 130590
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EEF1A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139487221] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EEF1A1
- Gene Name
- eukaryotic translation elongation factor 1 alpha 1
View Full Product Details