Product Details

SNP ID
rs140600321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143846071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGATAGACAATGAAGAAAGGCG[A/G]GCAGAACAGAGGAAGTATGGAGTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LTV1 PubMed Links

Gene Details

Gene
LTV1
Gene Name
LTV1 ribosome biogenesis factor
There are no transcripts associated with this gene.

View Full Product Details