Product Details
- SNP ID
-
rs140677601
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:116574584 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTTTTTGACAAATCTTTAATTCT[G/T]TCACTTTCACCACCATCTGTCCCCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RWDD1
PubMed Links
Gene Details
- Gene
- RWDD1
- Gene Name
- RWD domain containing 1
View Full Product Details