Product Details

SNP ID
rs140689285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57052925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATTGTTGGAATGAAAAACAAGC[A/G]TTTATGTTTACAGAACAATACAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1586 PubMed Links

Gene Details

Gene
KIAA1586
Gene Name
KIAA1586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286274.1 628 Silent Mutation GCA,GCG A115A NP_001273203.1
NM_001286275.1 628 Silent Mutation GCA,GCG A75A NP_001273204.1
NM_001286276.1 628 Silent Mutation GCA,GCG A75A NP_001273205.1
NM_020931.3 628 Silent Mutation GCA,GCG A142A NP_065982.1
XM_011514767.2 628 Silent Mutation GCA,GCG A142A XP_011513069.1

View Full Product Details