Product Details

SNP ID
rs140795393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:1314190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTCCCTTCATAAGGACTTTTT[C/T]TGTCTTTCTTTAACGCCCAGGCTTC
Phenotype
MIM: 612788
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXQ1 PubMed Links

Gene Details

Gene
FOXQ1
Gene Name
forkhead box Q1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033260.3 1751 UTR 3 NP_150285.3
Gene
LINC01394
Gene Name
long intergenic non-protein coding RNA 1394
There are no transcripts associated with this gene.

Gene
LOC107986554
Gene Name
uncharacterized LOC107986554
There are no transcripts associated with this gene.

View Full Product Details