Product Details

SNP ID
rs140859475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143501776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCAAAACATGCAAATTCTTCC[C/T]CTTCTCTTTATATTAGAGTAAGCAA
Phenotype
MIM: 136820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUCA2 PubMed Links

Gene Details

Gene
FUCA2
Gene Name
fucosidase, alpha-L- 2, plasma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032020.4 Intron NP_114409.2

View Full Product Details