Product Details

SNP ID
rs140910734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:146029933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGATGAGAAGGATGGGATCAACC[A/G]GTGTCTGCCTGACGGCCAGTCCCTC
Phenotype
MIM: 604473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRM1 PubMed Links

Gene Details

Gene
GRM1
Gene Name
glutamate metabotropic receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278064.1 895 Missense Mutation CAG,CGG Q139R NP_001264993.1
NM_001278065.1 895 Missense Mutation CAG,CGG Q139R NP_001264994.1
NM_001278066.1 895 Missense Mutation CAG,CGG Q139R NP_001264995.1
NM_001278067.1 895 Missense Mutation CAG,CGG Q139R NP_001264996.1
XM_011535782.1 895 Missense Mutation CAG,CGG Q139R XP_011534084.1
XM_017010783.1 895 Missense Mutation CAG,CGG Q139R XP_016866272.1
XM_017010784.1 895 Missense Mutation CAG,CGG Q139R XP_016866273.1
XM_017010785.1 895 Missense Mutation CAG,CGG Q139R XP_016866274.1
XM_017010786.1 895 Missense Mutation CAG,CGG Q139R XP_016866275.1
XM_017010787.1 895 Missense Mutation CAG,CGG Q139R XP_016866276.1
XM_017010788.1 895 Intron XP_016866277.1

View Full Product Details