Product Details
- SNP ID
-
rs141418706
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:22569653 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTAAAGGGCTATGCCCACTGGCC[A/G]GCGAGGATAGAGCACATGACCCAGC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HDGFL1
PubMed Links
Gene Details
- Gene
- HDGFL1
- Gene Name
- hepatoma derived growth factor-like 1
There are no transcripts associated with this gene.
View Full Product Details