Product Details

SNP ID
rs141424762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2766417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCAACGAAATCCCCTCGCTTAT[C/T]CTGTGGGGGCCGCCGGGCTGCGGCA
Phenotype
MIM: 608196
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYLK4 PubMed Links

Gene Details

Gene
MYLK4
Gene Name
myosin light chain kinase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249078.3 986 Intron XP_005249135.2
XM_005249079.2 986 Intron XP_005249136.1
XM_006715082.3 986 Intron XP_006715145.1
XM_011514567.2 986 Intron XP_011512869.1
XM_017010819.1 986 Intron XP_016866308.1
Gene
WRNIP1
Gene Name
Werner helicase interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020135.2 986 Silent Mutation ATC,ATT I265I NP_064520.2
NM_130395.2 986 Silent Mutation ATC,ATT I265I NP_569079.1
XM_005249232.3 986 Silent Mutation ATC,ATT I265I XP_005249289.1
XM_017011060.1 986 Silent Mutation ATC,ATT I265I XP_016866549.1

View Full Product Details