Product Details

SNP ID
rs141495600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73466357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACCGGCGTAAGGGACCAGCTGT[A/G]TGGGGTCTGAGAGCTCAGATTGATA
Phenotype
MIM: 614667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTO1 PubMed Links

Gene Details

Gene
MTO1
Gene Name
mitochondrial tRNA translation optimization 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123226.1 490 Silent Mutation GTA,GTG V122V NP_001116698.1
NM_012123.3 490 Silent Mutation GTA,GTG V122V NP_036255.2
NM_133645.2 490 Silent Mutation GTA,GTG V122V NP_598400.1
XM_006715444.2 490 Silent Mutation GTA,GTG V48V XP_006715507.1
XM_006715445.2 490 Silent Mutation GTA,GTG V48V XP_006715508.1
XM_006715446.3 490 Silent Mutation GTA,GTG V122V XP_006715509.1
XM_011535723.1 490 UTR 5 XP_011534025.1
XM_011535724.1 490 UTR 5 XP_011534026.1
XM_011535725.2 490 UTR 5 XP_011534027.1
XM_017010697.1 490 Silent Mutation GTA,GTG V48V XP_016866186.1
XM_017010698.1 490 UTR 5 XP_016866187.1
XM_017010699.1 490 UTR 5 XP_016866188.1
XM_017010700.1 490 UTR 5 XP_016866189.1

View Full Product Details