Product Details

SNP ID
rs141511769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:93242699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTTTTTTCATTGAAAAGATAGT[A/G]TGTTGTCTGATCTTTACAAAAAAAT
Phenotype
MIM: 602190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHA7 PubMed Links

Gene Details

Gene
EPHA7
Gene Name
EPH receptor A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288629.1 3965 UTR 3 NP_001275558.1
NM_001288630.1 3965 Intron NP_001275559.1
NM_004440.3 3965 UTR 3 NP_004431.1
XM_005248669.2 3965 UTR 3 XP_005248726.1
XM_005248671.2 3965 Intron XP_005248728.1
XM_017010365.1 3965 UTR 3 XP_016865854.1
XM_017010366.1 3965 Intron XP_016865855.1

View Full Product Details