Product Details

SNP ID
rs141591240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:100510080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCTTTTGATTTGGGAAATCGAG[A/G]AGTAACTGCACAGCTCTCTGGCTTT
Phenotype
MIM: 614217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASCC3 PubMed Links

Gene Details

Gene
ASCC3
Gene Name
activating signal cointegrator 1 complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284271.1 6922 Intron NP_001271200.1
NM_006828.3 6922 Missense Mutation CCT,TCT P2105S NP_006819.2
NM_022091.4 6922 Intron NP_071374.1
XM_011535394.2 6922 Missense Mutation CCT,TCT P2110S XP_011533696.1
XM_011535395.2 6922 Missense Mutation CCT,TCT P2007S XP_011533697.1
XM_011535396.2 6922 Missense Mutation CCT,TCT P2007S XP_011533698.1
XM_017010205.1 6922 Missense Mutation CCT,TCT P2007S XP_016865694.1
XM_017010206.1 6922 Missense Mutation CCT,TCT P1632S XP_016865695.1

View Full Product Details