Product Details

SNP ID
rs141748507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27810193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCGGCGGTGTCAAGAAGCCCCA[C/T]CGCTATCGGCCTGGTACAGTGGCTC
Phenotype
MIM: 602787 MIM: 602791 MIM: 602800 MIM: 602802 MIM: 602818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AI PubMed Links

Gene Details

Gene
HIST1H2AI
Gene Name
histone cluster 1, H2ai
There are no transcripts associated with this gene.

Gene
HIST1H2AJ
Gene Name
histone cluster 1, H2aj
There are no transcripts associated with this gene.

Gene
HIST1H2BL
Gene Name
histone cluster 1, H2bl
There are no transcripts associated with this gene.

Gene
HIST1H2BM
Gene Name
histone cluster 1, H2bm
There are no transcripts associated with this gene.

Gene
HIST1H3H
Gene Name
histone cluster 1, H3h
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003536.2 Intron NP_003527.1

View Full Product Details