Product Details

SNP ID
rs141753924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:105281811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAATCAGTGGTCCAAGCATGGCC[A/G]ATGGTATATTTATGAAACTTCAGCA
Phenotype
MIM: 600400
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PREP PubMed Links

Gene Details

Gene
PREP
Gene Name
prolyl endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002726.4 2006 Silent Mutation ATC,ATT I591I NP_002717.3
XM_005267044.2 2006 Intron XP_005267101.1
XM_011535925.2 2006 Intron XP_011534227.1

View Full Product Details