Product Details

SNP ID
rs141784444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33277620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCAGGAAGCTTGCAGTGGTCC[C/T]GGGGCCCCTCCCTTCCTGCTCATCC
Phenotype
MIM: 603095 MIM: 180473 MIM: 603443 MIM: 611440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GALT4 PubMed Links

Gene Details

Gene
B3GALT4
Gene Name
beta-1,3-galactosyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003782.3 481 Silent Mutation CCC,CCT P67P NP_003773.1
Gene
MIR6873
Gene Name
microRNA 6873
There are no transcripts associated with this gene.

Gene
RPS18
Gene Name
ribosomal protein S18
There are no transcripts associated with this gene.

Gene
VPS52
Gene Name
VPS52, GARP complex subunit
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164267.1 481 Intron NP_001157739.1
NM_005452.5 481 Intron NP_005443.3

View Full Product Details