Product Details

SNP ID
rs141849753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32166601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCTGACCTTGTGCGCTGGGAGG[C/T]GCATCTGCAGCACTTACAGGTGAGG
Phenotype
MIM: 603099 MIM: 609897 MIM: 603298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGPAT1 PubMed Links

Gene Details

Gene
AGPAT1
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
There are no transcripts associated with this gene.

Gene
EGFL8
Gene Name
EGF like domain multiple 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030652.3 286 Missense Mutation CGC,TGC R69C NP_085155.1
Gene
MIR6721
Gene Name
microRNA 6721
There are no transcripts associated with this gene.

Gene
PPT2
Gene Name
palmitoyl-protein thioesterase 2
There are no transcripts associated with this gene.

Gene
PPT2-EGFL8
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details