Product Details

SNP ID
rs142261504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43507272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTCAGCCAACAGACTGTCCCCT[A/G]GTCCTGACGTCATTAGCACCATCTG
Phenotype
MIM: 610060 MIM: 612658 MIM: 609775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC73 PubMed Links

Gene Details

Gene
LRRC73
Gene Name
leucine rich repeat containing 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012974.2 783 Missense Mutation CCA,CTA P306L NP_001012992.1
NM_001271882.1 783 Missense Mutation CCA,CTA P183L NP_001258811.1
Gene
POLR1C
Gene Name
RNA polymerase I subunit C
There are no transcripts associated with this gene.

Gene
TJAP1
Gene Name
tight junction associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146016.1 783 Intron NP_001139488.1
NM_001146017.1 783 Intron NP_001139489.1
NM_001146018.1 783 Intron NP_001139490.1
NM_001146019.1 783 Intron NP_001139491.1
NM_001146020.1 783 Intron NP_001139492.1
NM_080604.2 783 Intron NP_542171.2
XM_006715249.1 783 Intron XP_006715312.1
XM_006715250.3 783 Intron XP_006715313.1
XM_006715251.2 783 Intron XP_006715314.1
XM_006715252.2 783 Intron XP_006715315.1
XM_006715254.3 783 Intron XP_006715317.1
XM_006715256.1 783 Intron XP_006715319.1
XM_006715257.2 783 Intron XP_006715320.1
XM_006715259.3 783 Intron XP_006715322.1
XM_006715260.1 783 Intron XP_006715323.1
XM_006715261.1 783 Intron XP_006715324.1
XM_006715262.2 783 Intron XP_006715325.1
XM_006715263.2 783 Intron XP_006715326.1
XM_006715265.1 783 Intron XP_006715328.1
XM_006715266.1 783 Intron XP_006715329.1
XM_006715267.2 783 Intron XP_006715330.1
XM_006715268.2 783 Intron XP_006715331.1
XM_006715269.2 783 Intron XP_006715332.1
XM_006715270.1 783 Intron XP_006715333.1
XM_011514995.1 783 Intron XP_011513297.1
XM_011514996.1 783 Intron XP_011513298.1
XM_017011488.1 783 Intron XP_016866977.1
XM_017011489.1 783 Intron XP_016866978.1
XM_017011490.1 783 Intron XP_016866979.1
XM_017011491.1 783 Intron XP_016866980.1
XM_017011492.1 783 Intron XP_016866981.1
XM_017011493.1 783 Intron XP_016866982.1
Gene
YIPF3
Gene Name
Yip1 domain family member 3
There are no transcripts associated with this gene.

View Full Product Details