Product Details

SNP ID
rs142270062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108044801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTAGGTCTCAGTTTGAATTTTCC[G/T]GAATATTTGCAAAACTGGTACTGGA
Phenotype
MIM: 607649
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OSTM1 PubMed Links

Gene Details

Gene
OSTM1
Gene Name
osteopetrosis associated transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014028.3 1075 Missense Mutation CAG,CCG Q330P NP_054747.2

View Full Product Details