Product Details

SNP ID
rs142359891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33423493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTCCAGATGTACGGGGACCCTC[C/T]ATGCACCGAACCCAATACGTTCATT
Phenotype
MIM: 616953 MIM: 602881 MIM: 603384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUTA PubMed Links

Gene Details

Gene
CUTA
Gene Name
cutA divalent cation tolerance homolog (E. coli)
There are no transcripts associated with this gene.

Gene
PHF1
Gene Name
PHD finger protein 1
There are no transcripts associated with this gene.

Gene
SYNGAP1
Gene Name
synaptic Ras GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006772.2 279 Silent Mutation TCC,TCT S28S NP_006763.2

View Full Product Details