Product Details

SNP ID
rs142398173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36598910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGGTGAAAAAAGAAGTAGAAAT[C/T]GTGGCCCACCTCCCTCTTGGGGTCG
Phenotype
MIM: 603364
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRSF3 PubMed Links

Gene Details

Gene
SRSF3
Gene Name
serine and arginine rich splicing factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003017.4 439 Missense Mutation CGT,TGT R90C NP_003008.1

View Full Product Details