Product Details

SNP ID
rs142400023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:131641527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGGACTCAGGAAACTGGAAAAG[A/C]AAGGTATACCCCTCAGCCTTTTCTC
Phenotype
MIM: 602182
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ENPP3 PubMed Links

Gene Details

Gene
ENPP3
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005021.4 301 Missense Mutation AAA,CAA K51Q NP_005012.2
XM_011535897.1 301 Intron XP_011534199.1
XM_017010932.1 301 Intron XP_016866421.1

View Full Product Details