Product Details

SNP ID
rs142429317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108561784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGTGGATGGTGCGTTGCGTGCC[C/G]TACTTCAAGGATAAGGGCGACAGCA
Phenotype
MIM: 602681
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FOXO3 PubMed Links

Gene Details

Gene
FOXO3
Gene Name
forkhead box O3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001455.3 Intron NP_001446.1
NM_201559.2 Intron NP_963853.1
XM_005266867.4 Intron XP_005266924.1
XM_005266868.3 Intron XP_005266925.1
XM_011535626.1 Intron XP_011533928.1
XM_011535628.2 Intron XP_011533930.1
XM_011535629.2 Intron XP_011533931.1
XM_017010585.1 Intron XP_016866074.1
XM_017010586.1 Intron XP_016866075.1

View Full Product Details