Product Details

SNP ID
rs142431486
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:96798492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTCACACCACCGTCCGATGTT[C/T]CCCACACACATAGACAGCACTAGGA
Phenotype
MIM: 606915
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GPR63 PubMed Links

Gene Details

Gene
GPR63
Gene Name
G protein-coupled receptor 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143957.2 2128 Missense Mutation AAA,GAA K414E NP_001137429.1
NM_030784.3 2128 Missense Mutation AAA,GAA K414E NP_110411.1
XM_006715570.3 2128 Missense Mutation AAA,GAA K414E XP_006715633.1
XM_011536153.2 2128 Missense Mutation AAA,GAA K414E XP_011534455.1
XM_011536154.2 2128 Missense Mutation AAA,GAA K414E XP_011534456.1
XM_011536155.2 2128 Missense Mutation AAA,GAA K414E XP_011534457.1
XM_011536157.2 2128 Missense Mutation AAA,GAA K414E XP_011534459.1
XM_011536159.2 2128 Missense Mutation AAA,GAA K414E XP_011534461.1
XM_017011334.1 2128 Missense Mutation AAA,GAA K414E XP_016866823.1

View Full Product Details