Product Details

SNP ID
rs142447236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133991209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCTACCACACAGCTTGTTACA[C/T]TCCAAGAGCTGCTCCTGGGGTTTTC
Phenotype
MIM: 610372 MIM: 605521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC2A12 PubMed Links

Gene Details

Gene
SLC2A12
Gene Name
solute carrier family 2 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145176.2 1971 Silent Mutation GAA,GAG E600E NP_660159.1
XM_006715349.3 1971 Intron XP_006715412.1
XM_017010311.1 1971 Intron XP_016865800.1
Gene
TBPL1
Gene Name
TATA-box binding protein like 1
There are no transcripts associated with this gene.

View Full Product Details