Product Details
- SNP ID
-
rs142456140
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:43432172 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGATTACATCCTACCCTGCAGTCC[A/T]GGATGGCGCCTCCGACTTGCAGCTT
- Phenotype
-
MIM: 612509
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ABCC10
PubMed Links
Gene Details
- Gene
- ABCC10
- Gene Name
- ATP binding cassette subfamily C member 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001198934.1 |
407 |
Silent Mutation |
CCA,CCT |
P64P |
NP_001185863.1 |
NM_033450.2 |
407 |
Silent Mutation |
CCA,CCT |
P21P |
NP_258261.2 |
XM_011514974.2 |
407 |
Silent Mutation |
CCA,CCT |
P115P |
XP_011513276.2 |
XM_011514983.2 |
407 |
Intron |
|
|
XP_011513285.1 |
XM_011514985.2 |
407 |
Intron |
|
|
XP_011513287.1 |
XM_011514986.2 |
407 |
Intron |
|
|
XP_011513288.1 |
XM_017011445.1 |
407 |
Silent Mutation |
CCA,CCT |
P115P |
XP_016866934.1 |
XM_017011446.1 |
407 |
Silent Mutation |
CCA,CCT |
P64P |
XP_016866935.1 |
XM_017011447.1 |
407 |
Silent Mutation |
CCA,CCT |
P64P |
XP_016866936.1 |
XM_017011448.1 |
407 |
Silent Mutation |
CCA,CCT |
P115P |
XP_016866937.1 |
XM_017011449.1 |
407 |
Silent Mutation |
CCA,CCT |
P115P |
XP_016866938.1 |
XM_017011450.1 |
407 |
Silent Mutation |
CCA,CCT |
P115P |
XP_016866939.1 |
XM_017011451.1 |
407 |
Silent Mutation |
CCA,CCT |
P115P |
XP_016866940.1 |
XM_017011452.1 |
407 |
UTR 5 |
|
|
XP_016866941.1 |
XM_017011453.1 |
407 |
Silent Mutation |
CCA,CCT |
P115P |
XP_016866942.1 |
XM_017011454.1 |
407 |
Intron |
|
|
XP_016866943.1 |
XM_017011455.1 |
407 |
Intron |
|
|
XP_016866944.1 |
XM_017011456.1 |
407 |
Intron |
|
|
XP_016866945.1 |
- Gene
- MIR6780B
- Gene Name
- microRNA 6780b
There are no transcripts associated with this gene.
View Full Product Details