Product Details

SNP ID
rs142666082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26598068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTACACAACACGCCTATGGGTGC[C/T]CGCAGGCGCAGCCCCTTCCGTTATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABT1 PubMed Links

Gene Details

Gene
ABT1
Gene Name
activator of basal transcription 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013375.3 436 Silent Mutation GCC,GCT A132A NP_037507.1
Gene
LOC105374989
Gene Name
uncharacterized LOC105374989
There are no transcripts associated with this gene.

View Full Product Details