Product Details

SNP ID
rs142672835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43306257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGTCAGGGTCTTGTGGCAAC[A/G]CTGGCACCTCAGACACGGTCGGTGC
Phenotype
MIM: 604995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRIP3 PubMed Links

Gene Details

Gene
CRIP3
Gene Name
cysteine rich protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206922.2 457 Missense Mutation NP_996805.2
XM_005249103.3 457 Missense Mutation XP_005249160.1
XM_011514609.2 457 Missense Mutation XP_011512911.1
XM_017010856.1 457 Missense Mutation XP_016866345.1
XM_017010857.1 457 Missense Mutation XP_016866346.1
Gene
SLC22A7
Gene Name
solute carrier family 22 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006672.3 457 Intron NP_006663.2
NM_153320.2 457 Intron NP_696961.2
XM_006714970.3 457 Intron XP_006715033.1
XM_006714971.3 457 Intron XP_006715034.1
XM_011514256.2 457 Intron XP_011512558.1
XM_011514257.2 457 Intron XP_011512559.1
XM_011514259.1 457 Intron XP_011512561.1
XM_011514261.2 457 Intron XP_011512563.1
XM_011514262.2 457 Intron XP_011512564.1
XM_011514263.2 457 Intron XP_011512565.1
XM_017010198.1 457 Intron XP_016865687.1
XM_017010199.1 457 Intron XP_016865688.1
XM_017010200.1 457 Intron XP_016865689.1
XM_017010201.1 457 Intron XP_016865690.1

View Full Product Details