Product Details

SNP ID
rs142692484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:106629702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCATGCTGGGCCGGAGCCTCCG[A/C]GAAGTGAGTGGAATTGGCCCGCTGA
Phenotype
MIM: 610502
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
QRSL1 PubMed Links

Gene Details

Gene
QRSL1
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018292.4 125 Silent Mutation CGA,CGC R7R NP_060762.3
XM_011535924.2 125 UTR 5 XP_011534226.1
Gene
RTN4IP1
Gene Name
reticulon 4 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318746.1 125 Intron NP_001305675.1
NM_032730.5 125 Intron NP_116119.2
XM_011536192.2 125 Intron XP_011534494.1
XM_017011375.1 125 Intron XP_016866864.1
XM_017011376.1 125 Intron XP_016866865.1

View Full Product Details