Product Details

SNP ID
rs142757005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:122998039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCAGAACCTGGACATGTTCAAA[A/C]GCTTTAAGGCCACCGACCCTGGCAT
Phenotype
MIM: 616945
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLVS2 PubMed Links

Gene Details

Gene
CLVS2
Gene Name
clavesin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010852.3 3299 Missense Mutation AGC,CGC S88R NP_001010852.2
XM_017010266.1 3299 Missense Mutation AGC,CGC S88R XP_016865755.1

View Full Product Details