Product Details

SNP ID
rs142811934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149595125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAAACCTTTTTTAAAGCCATCT[C/T]GAAATCCTCCATAGTTGTAGGCATG
Phenotype
MIM: 606696
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GINM1 PubMed Links

Gene Details

Gene
GINM1
Gene Name
glycoprotein integral membrane 1
There are no transcripts associated with this gene.

Gene
KATNA1
Gene Name
katanin catalytic subunit A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204076.1 1736 UTR 3 NP_001191005.1
NM_007044.3 1736 Missense Mutation AAG,GAG K463E NP_008975.1
XM_005266804.2 1736 Missense Mutation AAG,GAG K463E XP_005266861.1
XM_005266805.3 1736 Missense Mutation AAG,GAG K410E XP_005266862.1
XM_017010207.1 1736 Missense Mutation AAG,GAG K463E XP_016865696.1
XM_017010208.1 1736 Missense Mutation AAG,GAG K335E XP_016865697.1
XM_017010209.1 1736 Missense Mutation AAG,GAG K335E XP_016865698.1
XM_017010210.1 1736 Missense Mutation AAG,GAG K463E XP_016865699.1
Gene
RPS18P9
Gene Name
ribosomal protein S18 pseudogene 9
There are no transcripts associated with this gene.

View Full Product Details