Product Details

SNP ID
rs142886458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:154406538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCTCGGATGTAGTCCGCACCC[C/T]GGACCAGATGCCGCTCGGTCGTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNKSR3 PubMed Links

Gene Details

Gene
CNKSR3
Gene Name
CNKSR family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173515.2 4472 Missense Mutation CAG,CGG Q495R NP_775786.2
XM_011535485.2 4472 Missense Mutation CAG,CGG Q485R XP_011533787.1

View Full Product Details