Product Details

SNP ID
rs142895740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39299491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGATGCTGTATGATTCCCATG[G/T]GTCCCTCTGGATAGCATCCTTGCTG
Phenotype
MIM: 607370
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNK17 PubMed Links

Gene Details

Gene
KCNK17
Gene Name
potassium two pore domain channel subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135111.1 1227 UTR 3 NP_001128583.1
NM_031460.3 1227 Missense Mutation CAC,CCC H312P NP_113648.2

View Full Product Details