Product Details

SNP ID
rs143467246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34247117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCAGCTGGGTCATAGCTGTAC[A/G]TGGGGAGCAGGTGATGGCGCAGCCG
Phenotype
MIM: 611419 MIM: 600701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf1 PubMed Links

Gene Details

Gene
C6orf1
Gene Name
chromosome 6 open reading frame 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008703.2 625 Missense Mutation CGT,TGT R18C NP_001008703.1
NM_001008704.2 625 Missense Mutation CGT,TGT R18C NP_001008704.1
NM_001287396.1 625 Missense Mutation CGT,TGT R18C NP_001274325.1
NM_001287397.1 625 UTR 5 NP_001274326.1
NM_001287398.1 625 UTR 5 NP_001274327.1
NM_178508.4 625 Missense Mutation CGT,TGT R18C NP_848603.2
Gene
HMGA1
Gene Name
high mobility group AT-hook 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319077.1 625 Intron NP_001306006.1
NM_001319078.1 625 Intron NP_001306007.1
NM_001319079.1 625 Intron NP_001306008.1
NM_001319080.1 625 Intron NP_001306009.1
NM_001319081.1 625 Intron NP_001306010.1
NM_001319082.1 625 Intron NP_001306011.1
NM_002131.3 625 Intron NP_002122.1
NM_145899.2 625 Intron NP_665906.1
NM_145901.2 625 Intron NP_665908.1
NM_145902.2 625 Intron NP_665909.1
NM_145903.2 625 Intron NP_665910.1
NM_145905.2 625 Intron NP_665912.1
Gene
MIR6835
Gene Name
microRNA 6835
There are no transcripts associated with this gene.

View Full Product Details