Product Details

SNP ID
rs143564858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39299363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTAGTCTTTAGTTTGGGTGGA[C/G]ATGTGCAAAGCTTAAAATCAGGGGT
Phenotype
MIM: 607370
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNK17 PubMed Links

Gene Details

Gene
KCNK17
Gene Name
potassium two pore domain channel subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135111.1 1355 UTR 3 NP_001128583.1
NM_031460.3 1355 UTR 3 NP_113648.2

View Full Product Details