Product Details

SNP ID
rs143674381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109445281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCTCAATCTCATTTAGTCGCC[A/G]TTGGATGGTCTGAGGGGTACAAGAC
Phenotype
MIM: 607129 MIM: 603498
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159291.1 3145 Missense Mutation CGG,TGG R847W NP_001152763.1
NM_001286613.1 3145 Missense Mutation CGG,TGG R952W NP_001273542.1
NM_022765.3 3145 Missense Mutation CGG,TGG R933W NP_073602.3
Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
There are no transcripts associated with this gene.

Gene
SMPD2
Gene Name
sphingomyelin phosphodiesterase 2
There are no transcripts associated with this gene.

View Full Product Details