Product Details

SNP ID
rs143715803
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31707717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGAGGAAGATGCCGGGCGGTA[C/G]TGGTGCGCTGTGCTAGGTCAGCACC
Phenotype
MIM: 142620 MIM: 606038 MIM: 610437 MIM: 611404
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ABHD16A PubMed Links
Additional Information
For this assay, SNP(s) [rs9267547] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ABHD16A
Gene Name
abhydrolase domain containing 16A
There are no transcripts associated with this gene.

Gene
LY6G6D
Gene Name
lymphocyte antigen 6 complex, locus G6D
There are no transcripts associated with this gene.

Gene
LY6G6E
Gene Name
lymphocyte antigen 6 complex, locus G6E (pseudogene)
There are no transcripts associated with this gene.

Gene
LY6G6F
Gene Name
lymphocyte antigen 6 complex, locus G6F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003693.1 312 Nonsense Mutation TAC,TAG Y104* NP_001003693.1
Gene
MIR4646
Gene Name
microRNA 4646
There are no transcripts associated with this gene.

View Full Product Details